Learn More
Felodipine, Thermo Scientific™
A dihydropyridine calcium channel blocker
154.00 EUR - 383.00 EUR
Chemical Identifiers
| CAS | 72509-76-3 |
|---|---|
| Molecular Formula | C18H19Cl2NO4 |
| Molecular Weight (g/mol) | 384.253 |
| MDL Number | MFCD00868316 |
| InChI Key | RZTAMFZIAATZDJ-UHFFFAOYSA-N |
| Synonym | felodipine, plendil, flodil, munobal, renedil, dl-felodipine, perfudal, splendil, feloday, prevex |
| PubChem CID | 3333 |
| ChEBI | CHEBI:585948 |
| IUPAC Name | 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
| SMILES | CCOC(=O)C1=C(NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15443819
|
Thermo Scientific Alfa Aesar
J61195.MF |
50mg |
154.00 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15423819
|
Thermo Scientific Alfa Aesar
J61195.MC |
100mg |
221.00 EUR
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15433819
|
Thermo Scientific Alfa Aesar
J61195.MD |
383.00 EUR
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsFelodipine is used as a dihydropyridine calcium channel blocker. It displays high vascular selectivity; lowers arterial blood pressure without altering cardiac contractility. Antihypertensive.
Solubility
Soluble in DMSO. Insoluble in water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
| 72509-76-3 | |
| 384.253 | |
| RZTAMFZIAATZDJ-UHFFFAOYSA-N | |
| 3333 | |
| 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
| C18H19Cl2NO4 | |
| MFCD00868316 | |
| felodipine, plendil, flodil, munobal, renedil, dl-felodipine, perfudal, splendil, feloday, prevex | |
| CHEBI:585948 | |
| CCOC(=O)C1=C(NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C)C |
Specifications
| 72509-76-3 | |
| MFCD00868316 | |
| felodipine, plendil, flodil, munobal, renedil, dl-felodipine, perfudal, splendil, feloday, prevex | |
| RZTAMFZIAATZDJ-UHFFFAOYSA-N | |
| 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate | |
| 3333 | |
| 384.25 |
| C18H19Cl2NO4 | |
| 14,3953 | |
| Soluble in DMSO; Insoluble in water | |
| CCOC(=O)C1=C(NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C)C | |
| 384.253 | |
| CHEBI:585948 | |
| Felodipine |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'rtecsNumber' : US7968700
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.