missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO6 Monoclonal Antibody (OTI7C2), TrueMAB™, OriGene
Description
FBXO6 Monoclonal antibody specifically detects FBXO6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
| Antigen | FBXO6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI7C2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | FBXO6 |
| Gene Accession No. | Q9NRD1 |
| Gene Alias | AA408845; FBG1; Fbg2; F-box only protein 6; F-box only protein 6 b; F-box only protein 6b; F-box protein 6; F-box protein FBG2; F-box protein Fbx6; F-box protein that recognizes sugar chains 2; F-box/G-domain protein 2; Fbs1; FBS2; FBX2; FBX6; Fbx6b; FBXO6; Fbxo6b; NFB42; RP3-330O12.3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?