missing translation for 'onlineSavingsMsg'
Learn More

Favipiravir, TRC

Product Code. 17994655
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17994655 1 g 1g
17913115 10 mg 10mg
17932045 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994655 Supplier Toronto Research Chemicals Supplier No. TRCF1033501G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259793-96-9
Molecular Formula C5H4FN3O2
Molecular Weight (g/mol) 157.1
Synonym 2-Pyrazinecarboxamide, 6-fluoro-3,4-dihydro-3-oxo-, Pyrazinecarboxamide, 6-fluoro-3,4-dihydro-3-oxo- (9CI), 6-Fluoro-3,4-dihydro-3-oxo-2-pyrazinecarboxamide, 6-Fluoro-3-hydroxy-2-pyrazinecarboxamide, Avigan, Favipiravir, T 705, T 705 (pharmaceutical)
IUPAC Name 5-fluoro-2-oxo-1H-pyrazine-3-carboxamide
SMILES NC(=O)C1=NC(=CNC1=O)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H4FN3O2/c6-2-1-8-5(11)3(9-2)4(7)10/h1H,(H2,7,10)(H,8,11)
Formula Weight 157.029
Percent Purity >95%
Chemical Name or Material Favipiravir
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.