missing translation for 'onlineSavingsMsg'
Learn More

Favipiravir, MedChemExpress

Product Code. 16466158
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466158 10 mg 10mg
16448757 5 mg 5mg
16442297 50 mg 50mg
16436546 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466158 Supplier Medchem Express Supplier No. HY14768
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Favipiravir (T-705) is a potent viral RNA polymerase inhibitor, it is phosphoribosylated by cellular enzymes to its active form, Favipiravir-ribofuranosyl-5′-triphosphate (RTP). Favipiravir-RTP inhibits the influenza viral RNA-dependent RNA polymerase (RdRP) activity with an IC50 of 341 nM.

Favipiravir (T-705) is a potent viral RNA polymerase inhibitor, it is phosphoribosylated by cellular enzymes to its active form, Favipiravir-ribofuranosyl-5′-triphosphate (RTP). Favipiravir-RTP inhibits the influenza viral RNA-dependent RNA polymerase (RdRP) activity with an IC50 of 341 nM.

Specifications

Chemical Name or Material Favipiravir
Physical Form Solid
CAS 259793-96-9
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C5H4FN3O2
For Use With (Application) COVID-19-anti-virus
Synonym T-705
Solubility Information DMSO : 100 mg/mL (636.54 mM; Need ultrasonic) ∣H2O : 6.25 mg/mL (39.78 mM; Need ultrasonic)
SMILES O=C(N)C1=NC(F)=CNC1=O
Molecular Weight (g/mol) 157.1
Formula Weight 157.1
Percent Purity 98.5%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.