missing translation for 'onlineSavingsMsg'
Learn More

Fast garnet GBC base, MP Biomedicals™

Catalog Number 11408060
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 97-56-3
Molecular Formula C14H15N3
Synonym Oil yellow 170, Solvent yellow 3
InChI Key PFRYFZZSECNQOL-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C
IUPAC Name 2-methyl-4-[(2-methylphenyl)diazenyl]aniline
Molecular Weight (g/mol) 225.295
PubChem CID 7340
Formula Weight 225.3
Color Orange
Quantity 100 g
Physical Form Powder
Chemical Name or Material Fast garnet GBC base
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'