missing translation for 'onlineSavingsMsg'
Learn More

Fast Blue B Salt, MP Biomedicals™

Product Code. 11436652 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436652 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11436652 Supplier MP Biomedicals Supplier No. 0215020125
  • ONLINE EXCLUSIVE PRICE
    212.00 EUR
    201.60 EUR / 25g
    Save 10.40 EUR
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Used as a tissue stain. Fast Blue B Salt, MP Biomedicals is useful for TLC of Aflatoxins.

TRUSTED_SUSTAINABILITY

Specifications

CAS 14263-94-6
Molecular Formula C14H16Cl4N4O2Zn+2
Synonym Azoic Diazo No. 48, Diazo Blue B, o-Dianisidine bis(diazotized) zinc double salt, Naphthanil Diazo Blue B, DBB
Solubility Information Soluble in water (4mg/4mL, clear solution).
InChI Key ZZJFLKSMXLBSAQ-UHFFFAOYSA-N
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]#N)OC)[N+]#N.Cl.Cl.Cl.Cl.[Zn]
IUPAC Name 4-(4-diazonio-3-methoxyphenyl)-2-methoxybenzenediazonium;zinc;tetrahydrochloride
Molecular Weight (g/mol) 479.488
PubChem CID 129894309
Formula Weight 475.47
Quantity 25 g
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • Fast Blue B Salt
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: fast blue B salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.