missing translation for 'onlineSavingsMsg'
Learn More

Farnesol MP Biomedicals

Product Code. 11428050 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428050 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11428050 Supplier MP Biomedicals Supplier No. 0210168080

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4602-84-0
Density 0.886 g/mL
Molecular Formula C15H26O
MDL Number MFCD00002918,MFCD00002918,MFCD00002918,MFCD00002918
Quantity 100 mL
Synonym e,z-farnesol, 2-trans,6-cis-farnesol, 2e,6z-farnesol, trans,cis-farnesol, farnesol, 2e,6z-3,7,11-trimethyldodeca-2,6,10-trien-1-ol, farnesyl alcohol, e,z-3,7,11-trimethyl-2,6,10-dodecatrien-1-ol, 2-trans,6-cis-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
InChI Key CRDAMVZIKSXKFV-YFVJMOTDSA-N
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CO
IUPAC Name (2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
Molecular Weight (g/mol) 222.37
PubChem CID 1549109
ChEBI CHEBI:35966
Formula Weight 222.4
Chemical Name or Material Farnesol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.