missing translation for 'onlineSavingsMsg'
Learn More

Farnesol, 97.7%, MP Biomedicals™

Product Code. 11408050 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408050 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408050 Supplier MP Biomedicals Supplier No. 0210168005

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 4602-84-0
Color Colorless
Density 0.886g/mL
Assay Percent Range 97.7%
Molecular Formula C15H26O
MDL Number MFCD00002918,MFCD00002918,MFCD00002918,MFCD00002918
Quantity 5 mL
Synonym e,z-farnesol, 2-trans,6-cis-farnesol, 2e,6z-farnesol, trans,cis-farnesol, farnesol, 2e,6z-3,7,11-trimethyldodeca-2,6,10-trien-1-ol, farnesyl alcohol, e,z-3,7,11-trimethyl-2,6,10-dodecatrien-1-ol, 2-trans,6-cis-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
InChI Key CRDAMVZIKSXKFV-YFVJMOTDSA-N
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CO
IUPAC Name (2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
Molecular Weight (g/mol) 222.37
PubChem CID 1549109
ChEBI CHEBI:35966
Formula Weight 222.36
Physical Form Liquid
Chemical Name or Material Farnesol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.