Learn More
Kimberly-Clark™ Kimtech™ WETTASK™ Roll Wiper Dispenser
Description
Large, perforated roll for use in busy areas. Can be used with a range of mobile or mounted dispensing systems. Control the costs of solvents and disinfectants and improve safety by reducing spills.
Specifications
Specifications
| Product Type | Wipers |
| Length (Metric) | 32 cm |
| Width (Metric) | 31 cm |
| Material | Fiber |
| Color | White |
| For Use With (Application) | Wiping and preparing industrial and healthcare surfaces using your chosen solvent, chemical, detergent |
| Ply | 1-ply |