missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F-Spondin/SPON1 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB300-339-0.025mg
This item is not returnable.
View return policy
Description
F-Spondin/SPON1 Polyclonal specifically detects F-Spondin/SPON1 in Human, Mouse, Rat samples. It is validated for Western Blot, ICC/IF (Negative).
Specifications
| F-Spondin/SPON1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SPON1 | |
| Synthetic peptide, corresponding to amino acids 549-807 of SPON1. | |
| 0.025 mg | |
| Neuroscience | |
| 10418 | |
| Human |
| Western Blot | |
| 0.91 mg/mL | |
| Western Blot 1:200 - 1:500, ICC/IF (Negative) | |
| f-spondin, KIAA0762VSGP/F-spondin, MGC10724, spondin 1, (f-spondin) extracellular matrix protein, spondin 1, extracellular matrix protein, spondin-1, Vascular smooth muscle cell growth-promoting factor, VSGP | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| SPON1 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction