missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F-Spondin/SPON1 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
208.00 EUR - 370.00 EUR
Specifications
| Antigen | F-Spondin/SPON1 |
|---|---|
| Concentration | 0.91 mg/mL |
| Dilution | Western Blot 1:200 - 1:500, ICC/IF (Negative) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18515714
|
Novus Biologicals
NB300-339-0.02mg |
0.02 mg |
208.00 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18513123
|
Novus Biologicals
NB300-339-0.025mg |
0.025 mg |
370.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
F-Spondin/SPON1 Polyclonal specifically detects F-Spondin/SPON1 in Human, Mouse, Rat samples. It is validated for Western Blot, ICC/IF (Negative).Specifications
| F-Spondin/SPON1 | |
| Western Blot 1:200 - 1:500, ICC/IF (Negative) | |
| Polyclonal | |
| Chicken | |
| Neuroscience | |
| PBS with 0.02% Sodium Azide | |
| 10418 | |
| Synthetic peptide, corresponding to amino acids 549-807 of SPON1. | |
| Affinity Purified | |
| SPON1 |
| 0.91 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| f-spondin, KIAA0762VSGP/F-spondin, MGC10724, spondin 1, (f-spondin) extracellular matrix protein, spondin 1, extracellular matrix protein, spondin-1, Vascular smooth muscle cell growth-promoting factor, VSGP | |
| SPON1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title