Learn More
Sartorius F-10 (HAM) Nutrient Mixture with L-Glutamine
Product Code. 31908348 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31908348 100 mL 100mL
31908480 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31908348 Supplier Sartorius Supplier No. 010901B
 Restricted Item
In Stock
Add to basket
Add to basket
Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells.
  • F-10 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including primary rat astrocytes and epithelial cells.
Specifications
Form Liquid
Product Type Ham's F-10 Cell Culture Media
With Additives Glutamine, Sodium Bicarbonate
Without Additives None
Quantity 100 mL