Learn More
Beskrivelse
- Unique lock rail system allows panel door to be closed even when lockouts are in place
- Accommodates multiple locks, eliminating the need for hasps during group lockouts
- Lock rail design also keeps weight off of the breakers, which protects delicate electrical components from damage
- Lock rails are made of durable isoplast polymers with acrylic adhesive backing
- "No shackle" locks with EX Panel Loc keys are sold separately and are especially useful when you want lockout devices to be kept at the point of need
- A spacing gasket is also available for low profile panels
Tekniske data
Tekniske data
| Product Type | Lock Rail |
| Alternate Part Number | BREAKER BLOCKER RAISED LOCK RAIL 2 PACK |
| Color | Yellow |
| Material | Fiberglass-reinforced Nylon |
| Dimensions (L x W x H) | 4 x 0.8 x 0.7 x in. |
| Length (English) | 4 in. |
| Quantity | 2 Pk. |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?