Learn More
Sterile Eye Wash Bottle
Product Code. 12424940
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12424940 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12424940 Supplier Safety First Supplier No. E404PL1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Keep a Sterile Eye Wash Bottle set on hand as a first aid kit refill per the Health and Safety ACOP L74 regulation for contingency supplies in case of emergency.
The 1997 Approved Code of Practice states that 'Where tap water is not readily available for eye irrigation, at least 1L of sterile water or sterile normal saline 0.9% in sealed disposable containers should be provided'.
Specifications
Product Type Eyewash Bottle
Composition Contain a Sterile Solution of Sodium Chloride (0.9%)
For Use With (Equipment) Eyewash Station
Sterility Sterile
Volume (Metric) 500 mL