Learn More
MilliporeSigma™ EXtrelut™ NT Prepacked Columns
Product Code. 15893921
Change view
Click to view available options
Type:
EXtrelut NT 1 Prepacked Column, Glass
EXtrelut NT 20 Prepacked Column, Polyethylene
EXtrelut NT 3 Prepacked Column, Glass
Unit Size:
Pack of 100
Pack of 25
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15893921 EXtrelut NT 3 Prepacked Column, Glass Pack of 50
15883921 EXtrelut NT 1 Prepacked Column, Glass Pack of 100
15803931 EXtrelut NT 20 Prepacked Column, Polyethylene Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15893921 Supplier MilliporeSigma™ Supplier No. 1150950001
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
pre-packed columns for extraction of lipophilic compounds from aqueous solutions
Extrelut™ NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut™ NT1 can take up a maximum of one ml, Extrelut™ NT3 threemL and Extrelut™ NT20 up to twentymL of sample.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) To Obtain High Degree Purity
Sorbent SPE Sorbent
Type EXtrelut NT 3 Prepacked Column, Glass