Learn More
MilliporeSigma™ EXtrelut™ NT Prepacked Columns
Description
Extrelut™ NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut™ NT1 can take up a maximum of one ml, Extrelut™ NT3 threemL and Extrelut™ NT20 up to twentymL of sample.
Specifications
Specifications
| For Use With (Application) | To Obtain High Degree Purity |
| Sorbent | SPE Sorbent |
| Type | EXtrelut NT 3 Prepacked Column, Glass |