Learn More
Lenz Laborglasintrumente Extractors Acc. to Twisselmann
Product Code. 10375971
Change view
Click to view available options
Standard Taper Joint Bottom:
NS 29/32
NS 34/35
NS 45/40
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Bottom Unit Size
10375971 NS 45/40 Each
10223911 NS 29/32 Each
10283951 NS 34/35 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10375971 Supplier Lenz Laborglasintrumente Supplier No. 05380037
  • ONLINE EXCLUSIVE PRICE
    737.00 EUR
    603.52 EUR / Pack of 1
    Save 133.48 EUR
    18% Off
Estimated Shipment: 22-02-2027
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Extractor head with vacuum jacket
  • Made of DURAN™ tubing
  • Twisselmann condenser (improved) with thread GL 14
  • Round bottom flask
Specifications
Capacity (Metric) 100 mL
Material Duran Glass
Standard Taper Joint Bottom NS 45/40