Learn More
Lenz Laborglasintrumente Extractor Heads for Specific Light Solvents
Product Code. 10385971
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10385971 100 mL Each
10409752 250 mL Each
10624392 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10385971 Supplier Lenz Laborglasintrumente Supplier No. 05432137
  • ONLINE EXCLUSIVE PRICE
    270.00 EUR
    207.40 EUR / Pack of 1
    Save 62.60 EUR
    23% Off
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • For the extraction of liquids with specific light solvents
  • With insert filter (porosity: 0)
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 100 mL
Material Fritte P0
Standard Taper Joint NS 45/40