Learn More
Lenz Laborglasintrumente Extractor Heads Acc. To Twisselmann
Product Code. 10335691
Change view
Click to view available options
Capacity (Metric):
70 mL
100 mL
150 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10335691 70 mL Each
10421093 150 mL Each
10602862 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10335691 Supplier Lenz Laborglasintrumente Supplier No. 05415131
  • ONLINE EXCLUSIVE PRICE
    164.00 EUR
    125.21 EUR / Pack of 1
    Save 38.79 EUR
    23% Off
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Extractor heads
  • Acc. to Twisselmann
  • With vacuum jacket
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 70 mL
Material Borosilicate Glass 3.3
Quantity 1 Ea.
Standard Taper Joint NS 34/35