Learn More
Scotch-Brite™ Extra Thick Delicate Surface Cleaning Pads 350
Product Code. 17108830
Change view
Click to view available options
Dimensions (L x W):
158 x 95 mm
Unit Size:
Pack of 40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17108830 158 x 95 mm Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17108830 Supplier Scotch-Brite™ Supplier No. 7100180770
  • ONLINE EXCLUSIVE PRICE
    127.00 EUR
    77.03 EUR / Pack of 40
    Save 49.97 EUR
    39% Off
Estimated Shipment: 29-12-2026
Specifications
Product Type Cleaning Pad
Dimensions (L x W) 158 x 95 mm
Length (Metric) 158 mm
Quantity 40 Pk.
Style Extra Thick
Width (Metric) 95 mm