Learn More
Velp™ External Probe
Product Code. 15339654 Shop All Velp Products
Change view
Click to view available options
Diameter (Metric) Body:
3 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Body Unit Size
15339654 3 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15339654 Supplier Velp™ Supplier No. A00000268
  • ONLINE EXCLUSIVE PRICE
    93.00 EUR
    90.50 EUR / Pack of 1
    Save 2.50 EUR
    2% Off
Estimated Shipment: 01-12-2026
External Probe is for use with AREC.X and AREX digital hotplate stirrers
VELP External Probe is for use with AREC.X and AREX digital hotplate stirrers
  • Accessory
Specifications
Accuracy ± 1°C (PT100)
For Use With (Equipment) AREC.X and AREX DIGITAL
Material Stainless Steel
Temperature (Metric) Operating 310°C
Type Temperature Probe
Diameter (Metric) Body 3 mm