Learn More
Gibco™ Expi293™ Met (–) Protein Labeling Kit
Product Code. 17114719
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Product Code. 17114719
Brand: Gibco™ A41249
Expi293 Met (-) Protein Labeling Kit includes Expi293 Met (-) Expression Medium and the ExpiFectamine 293 Met (-) Transfection Kit for rapid, high-yield transient protein expression from Expi293F cells for applications where labeling of methionine residues in proteins is desired
Kit components are specifically formulated without methionine, allowing users to add labeled methionine of their choice
Features:
  • Methionine-free medium and transfection enhancer formulations
  • 2 to 10-fold higher protein yields compared to other transfection reagents used for high-density 293 cell culture
  • Same transient transfection protocol as ExpiFectamine 293 Transfection Kit
  • Scalable transfection from small to large scale while maintaining equivalent volumetric protein yields
Specifically designed to easily integrate with other components of the Expi293 Expression System, including Expi293F cells and Expi293 Expression Medium
Components included:
  • 1 L Expi293 Met (–) Expression Medium
  • 1 ExpiFectamine 293 Met (–) Transfection Kit sufficient to transfect 1 L of culture
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protein Labeling Kit
Quantity 1 kit
Sufficient For Protein Production
Content And Storage
  • 3 x 0.9 mL ExpiFectamine 293 Transfection Reagent, store at 2°C to 8°CC
  • 5 mL ExpiFectamine 293 Enhancer 1, store at 2°C to 8°CC
  • 50 mL ExpiFectamine 293 Met (–) Enhancer 2, store at 2°C to 8°CC
  • 100 mL Opti-Plex Complexation Buffer, store at 2°C to 8°CC (protect from light)
  • 1 L Expi293 Met (–) Expression Medium, store at 2°C to 8°CC (protect from light)
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Line Expi293F