missing translation for 'onlineSavingsMsg'
Learn More

Exemestane, MedChemExpress

Product Code. 16471937
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16471937

missing translation for 'mfr': Medchem Express HY13632

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Exemestane (FCE 24304) is a selective, irreversible and orally active steroidal aromatase inhibitor with IC50s of 30 nM and 40 nM for human placental and rat ovarian aromatase, respectively. Exemestane can be used for hormone-dependent breast cancer research.

Exemestane (FCE 24304) is a selective, irreversible and orally active steroidal aromatase inhibitor with IC50s of 30 nM and 40 nM for human placental and rat ovarian aromatase, respectively. Exemestane can be used for hormone-dependent breast cancer research.

Specifications

Chemical Name or Material Exemestane
Physical Form Solid
CAS 107868-30-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H24O2
For Use With (Application) Cancer-programmed cell death
Synonym FCE 24304 EXE
Solubility Information DMSO : ≥ 54 mg/mL (182.19 mM)
SMILES C[C@]1([C@](CC2)([H])[C@]3([H])CC(C4=CC(C=C[C@]4(C)[C@@]3([H])CC1)=O)=C)C2=O
Molecular Weight (g/mol) 296.4
Formula Weight 296.4
Percent Purity 99.92%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.