Learn More
Epredia™ Excelsior™ AS Tissue Processor Accessories
Product Code. 13470487 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Air System Duct Adapter Kit
Basket Carrying Container
Downdraft Extrctn Duct Adapter
Monitor Clamp Kit
Organized Basket, 222 cassette
Organized Basket, 300 cassette
Potassium Permanganate Filter
Storage Bottle
Vapor Filter (Charcoal)
Waste Wax Drawer Kit
Unit Size:
Each
Pack of 6
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
13470487 Vapor Filter (Charcoal) Pack of 6
12638156 Downdraft Extrctn Duct Adapter Each
12648156 Air System Duct Adapter Kit Each
12683156 Storage Bottle Pack of 6
12678156 Waste Wax Drawer Kit Each
12688156 Monitor Clamp Kit Each
12608346 Basket Carrying Container Each
12764856 Potassium Permanganate Filter Pack of 6
12658156 Organized Basket, 222 cassette Pack of 6
15594280 Organized Basket, 300 cassette Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 13470487 Supplier Epredia™ Supplier No. 7411258
  • ONLINE EXCLUSIVE PRICE
    687.00 EUR
    668.93 EUR / Pack of 6
    Save 18.07 EUR
    2% Off
In Stock
Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.
For use with Shandon Excelsior ES Tissue Processor
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vapor Filter (Charcoal)
For Use With (Equipment) Excelsior AS Tissue Processor
Quantity Cs. of 6 Filters