Learn More
R&D Systems™ ExCellerate Human NK Cell Expansion Media, Xeno-Free
Product Code. 18747856
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18747856 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18747856 Supplier R&D Systems™ Supplier No. CCM032
  • ONLINE EXCLUSIVE PRICE
    179.17 EUR
    158.55 EUR / 500mL
    Save 20.62 EUR
    11% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Media for the ex vivo expansion and culture of human Natural Killer lymphocytes under xeno-free conditions
TRUSTED_SUSTAINABILITY
Specifications
Product Line R&D Systems
Quantity 500 mL
Species Human, Multi-Species
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.