Description
- Exact volume transfer pipets are easy to use, economical and accurate. The pipets are produced from chemically resistant, low density polyethylene (LDPE) and are designed to dispense precise volumes. They are available in 5 popular sizes. Additional sizes are available on special order.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Bulb Draw (Metric) | 100 μL |
| Calibrated | No |
| Material | LDPE |
| Sterility | Non-sterile |
| Graduated | No |
| Length (English) | 3.3 in. |
| Packaging Type | 500/Pk., 10 Pk/Cs. |
| Color | Natural |
| Volume (Metric) | 0.10 mL |
| Show More |