missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EVOS™ Onstage Incubator (OSI-2) Master Plate

Product Code. 17826468
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17826468 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17826468 Supplier Invitrogen™ Supplier No. AMEPVH106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The EVOS Onstage Incubator (OSI-2) Master Plate is specifically designed to hold the OSI-2 environmental chamber when used with either the EVOS M5000 Imaging System or EVOS M7000 Imaging System.

The EVOS Onstage Incubator (OSI-2) Master Plate is specifically designed to hold the OSI-2 environmental chamber when used with either the EVOS M5000 Imaging System or EVOS M7000 Imaging System. The OSI-2 master plate is a component of the OSI-2 (link to new PDP page for AMC2000), offered separately in the event the stage plate needs replacing. Only vessel holders designed for the EVOS Onstage Incubator (link to vessel holder selection guide, currently being updated or online selection tool if the latter is available) should be used. This master plate, together with the OSI-2 vessel holder, accommodates various types of cell culture vessels and provides flexibility in your approach to extended time-lapse imaging experiments under precisely controlled environmental conditions.

TRUSTED_SUSTAINABILITY

Specifications

Type Onstage Accessory
Product Line EVOS
Description EVOS™ OSI-2 Master Plate
For Use With (Equipment) EVOS™ Onstage Incubator (OSI-2)
Holds Onstage incubator compatible vessel holders & vessel
Quantity 1 kit
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.