Learn More
Invitrogen™ EVOS™ Light Cube, Cy7 2.0
Product Code. 17468092
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17468092 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17468092 Supplier Invitrogen™ Supplier No. AMEP4967
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
The second-generation EVOS LED light cube provides superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality. At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through.
  • EVOS LED light cube provides superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality
  • Optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through
  • Condensing optics and hard-coated filters combined into one remarkable light engine that delivers digitally controlled fluorophore excitation
  • These LED light cubes are backwards compatible with older EVOS and Countess systems and can be used with first-generation light cubes
  • Like the original light cubes, these LED cubes are user-changeable and automatically recognized by the instrument
  • Ideal for use with Cy7 and Alexa Fluor 750, and other fluorophores with excitation and emission maxima near 716 and 794 nm
  • Stable illumination intensity over 50,000 hours helps experimental reproducibility
TRUSTED_SUSTAINABILITY
Specifications
Type LED Light Cube
Product Line EVOS
Excitation/Emission Ex: 716/40 Em: 794/32
For Use With (Equipment) Countess™ III FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System
Includes Condensing Optics, Hard-coated Filters
Quantity 1 each