Learn More
Thermo Scientific™ eVol™ XR Dispensing System Plungers
Artikelnummer. 15260369 Shop alle Thermo Scientific produkter
Klik for at se tilgængelige muligheder
For Use With (Equipment):
eVol System, 1000 μL
eVol System, 500 μL
Pakningsstørrelse:
Each
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15260369
Brand: Thermo Scientific™ 66002041
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Denne vare kan ikke returneres. Se returpolicy
Order these plungers for use with the respective size syringe on the Thermo Scientific™ eVol­™ Dispensing System.
Use these Thermo Scientific™ eVol™ Dispensing System Plungers with the respective size syringe on the eVol Dispensing System. Like the syringes, these plungers are available in 5 to 1000μL sizes. The unique, hand-held eVol Dispensing System accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
Exceptional Versatility and Functionality
  • Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
  • Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system
TRUSTED_SUSTAINABILITY
Tekniske data
Product Type Dispensing System Plunger
Product Line HyperSep
For Use With (Equipment) eVol System, 1000 μL
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.