Learn More
Evans blue, MP Biomedicals™
Product Code. 11416912 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
100 g
Unit Size:
100g
10g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416912 5 g 5g
11426912 10 g 10g
11436912 50 g 50g
11446912 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11416912 Supplier MP Biomedicals Supplier No. 0215110805
  • ONLINE EXCLUSIVE PRICE
    41.90 EUR
    39.90 EUR / 5g
    Save 2.00 EUR
    4% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Chemical Identifiers
CAS 314-13-6
Molecular Formula C34H24N6Na4O14S4
Molecular Weight (g/mol) 960.79
MDL Number MFCD00004021
InChI Key KBNIFDASRCWYGC-GXNXWABVSA-J
Synonym Direct blue 53
PubChem CID 24832074
IUPAC Name tetrasodium (6E)-4-amino-6-[2-(4'-{2-[(2E)-8-amino-1-oxo-5,7-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-5-oxo-5,6-dihydronaphthalene-1,3-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
Specifications
Color Black
Quantity 5 g
Physical Form Powder
Formula Weight 960.82
Percent Purity 95%
Grade Not Available
Chemical Name or Material Evans blue, Dye content: ∽85%