Learn More
Novus Biologicals™ EV SEC Columns
Shop All Bio Techne Products
Click to view available options
Quantity:
10 Columns
5 Columns
Unit Size:
Pack of 10
Pack of 5
Specifications
Specifications
| Quantity | 5 Columns |
| Content And Storage | Store at 4°C. |
| Sample Type | Biofluids or cell culture supernatant |