Learn More
Thermo Scientific™ Eutech™ Salinity (NaCl) Standard Solutions
Product Code. 11579124 Shop All Thermo Scientific Products
Change view
Click to view available options
Concentration:
25 ppt
5 ppt
Unit Size:
480mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration Unit Size
11579124 25 ppt 480mL
15601717 5 ppt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11579124 Supplier Thermo Scientific™ Supplier No. 01X211231
  • ONLINE EXCLUSIVE PRICE
    29.85 EUR
    28.80 EUR / 480mL
    Save 1.05 EUR
    3% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Eutech™ offers Salinity Standard Solutions for use with Salinity Meters.
Eutech™ offers a range of Salinity Standard Solutions for use with Salinity Meters. Salinity measurements are common in industries like agriculture, aquaculture, hydroponics, food, pools and spas where it is necessary to monitor the salt level constantly.
Features:
  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Description:
  • ECNACL5PPT: 5 ppt (parts per thousand) Nacl calibration solution
  • ECNACL25PPT: 25 ppt (parts per thousand) Nacl calibration solution
  • ECNACL45PPT: 45 ppt(parts per thousand) Nacl calibration solution
TRUSTED_SUSTAINABILITY
Specifications
Concentration 25 ppt
Description 25 ppt NaCl Calibration Solution
Type NaCl Calibration Solution