Learn More
Thermo Scientific™ Eutech™ Automatic Temperature Compensation (ATC) Probe ECPHWPTEM03J
Product Code. 12971688
Change view
Click to view available options
Length (Metric) Cable:
3 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable Unit Size
12971688 3 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12971688 Supplier Thermo Scientific™ Supplier No. 01X021820
  • ONLINE EXCLUSIVE PRICE
    127.00 EUR
    118.00 EUR / Pack of 1
    Save 9.00 EUR
    7% Off
Estimated Shipment: 03-11-2026
Eutech™ offers a wide range of temperature probes used in various educational, laboratory, and industrial applications.
Eutech™ offers a wide range of temperature probes used in various educational, laboratory, and industrial applications. Eutech™ Automatic Temperature Compensation (ATC) Probe ECPHWPTEM03J is used with the ph 600 series meters.
Features:
  • 3 meter cable length
TRUSTED_SUSTAINABILITY
Specifications
Description ATC Probe
For Use With (Equipment) Eutech pH 620, 610, 600, PC650, PD650, PCD650 meters
Type Probe/Electrode
Length (Metric) Cable 3 m