Learn More
IKA™ EUROSTAR 60 Digital Overhead Stirrer
Catalog No. 13470567 Alle ansehen IKA Produkte
Ansicht ändern
Click to view available options
Plug Type:
EU
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Packungsgröße
13470567 EU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Catalog No. 13470567 Lieferant IKA™ Lieferanten-Nr. 0004446000
Estimated Shipment: 14-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Universally used strong laboratory stirrer for quantities up to 40 L (H2O). It automatically adjusts the speed through microprocessor controlled technology within the speed range.
  • Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions
  • Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically
  • Guarantees a constant speed even with changes in viscosities of the sample
  • Digital speed display
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection
  • Short-term overload operation
  • Slim casing
  • Quiet operation
  • Error code display
TRUSTED_SUSTAINABILITY
Spezifikation
Speed 30 to 2000 rpm
Capacity 40 L
Display 7 Segment LED
Voltage 230 V
Wattage 176 w
Height (Metric) 248 mm
Width (Metric) 86 mm
Electrical Requirements 230V, 50/60Hz, 176w
Frequency 50/60 Hz
Plug Type EU
Chuck Size 0.5 to 10 mm
Material Alu-Cast Coating/Thermoplastic Polymer
Model EUROSTAR 60 Digital
Motor Type Brushless DC
Temperature Range (Metric) 5°C to 40°C
Torque 60 Ncm
Viscosity 50,000 mPas
Mehr anzeigen Weniger anzeigen