Learn More
Gibertini Elettronica™ EU-C Atex Technical Balances
Product Code. 15593876
Encompass
Change view
Click to view available options
Capacity:
≥2 kg
10.2 kg
15 kg
4.2 kg
7.5 kg
950, 7500 g
Readability:
0.01 g
0.01, 0.1 g
0.1 g
0.5 g
Model:
EU-C 10000PTX
EU-C 15000 X
EU-C 2002 X
EU-C 4200 X
EU-C 7500 DRX
EU-C 7500 PTX
EU-C 7500PQX
Measurement Time:
1 sec.
2 sec.
2.5 sec.
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Readability Model Measurement Time unitSize
15593876 15 kg 0.5 g EU-C 15000 X 2 sec. Each
15583876 10.2 kg 0.1 g EU-C 10000PTX 2 sec. Each
15533886 7.5 kg 0.1 g EU-C 7500 PTX 2 sec. Each
15523886 950, 7500 g 0.01, 0.1 g EU-C 7500 DRX 1 sec. Each
15503886 ≥2 kg 0.01 g EU-C 2002 X 2.5 sec. Each
15513886 4.2 kg 0.01 g EU-C 4200 X 2.5 sec. Each
15307796 7.5 kg 0.1 g EU-C 7500PQX 2 sec. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15593876 Supplier Gibertini Elettronica™ Supplier No. EUC 10000 PTX
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Designed for use in hazardous environments with the risk of explosion, these scales are classified as ZONE 2. Gibertini Elettronica™ EU-C Atex Technical Balances are built with method of protection nAL for gas group IIC and temperature class T4 (135°C) according to regulation EEC 94/9.
  • LCD display with small-size decimal digits, for easier and more immediate reading
  • Waterproof and acid resistant membrane keyboard. Easy to use with the ON/OFF, TARE, MODE and PRINT keys (RANGE/PRINT for DR model)
  • Indication of the reached stable weight
  • Bar-graph indicator of dosage and remaining capacity of the balance
  • Parameters configurable by menu: reading in g (grams), lb (pound), oz (ounce), ct (carats), pcs (pieces), % (percentage)
  • Automatic end of range calibration (with external mass supplied on request)
  • Memory of the current weight in case of power failure
  • Enhanced anti-shock system
  • Metallic body with epoxide protection
  • Load cell nearly unbreakable and without transportation problems
  • Data output: I/O RS232 adjustable
  • 1, 2 and 5kg masses in class F1 for calibration and special executions available upon request
Specifications
Certifications/Compliance Classified as ZONE 2. Built with method of protect
Capacity 15 kg
Readability 0.5 g
For Use With (Application) Designed for use in hazardous environments with th
Linearity ±0.5 g
Material Metallic Body with Epoxide Protection
Measurement Time 2 sec.
Model EU-C 15000 X
Repeatability ±0.5 g
Temperature Range (Metric) 10°C to 40°C
Type Technical Balance
Dimensions (D x W x H) 255 x 260 x 120 mm
Width (Metric) Pan 260 mm
Depth (Metric) Pan 260 mm
Weight (Metric) 6 kg
Electrical Requirements 240 VAC
Voltage 240 VAC