missing translation for 'onlineSavingsMsg'
Learn More

Etoxazole, TRC

Product Code. 30011725
Change view
Click to view available options
Quantity:
100 mg
500 mg
25 g
Unit Size:
100mg
25g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011725 25 g 25g
17943205 500 mg 500mg
17954535 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011725 Supplier Toronto Research Chemicals Supplier No. TRCE93660025G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153233-91-1
Molecular Formula C21 H23 F2 N O2
Molecular Weight (g/mol) 359.41
Synonym Oxazole, 2-(2,6-difluorophenyl)-4-[4-(1,1-dimethylethyl)-2-ethoxyphenyl]-4,5-dihydro-, 2-(2,6-Difluorophenyl)-4-[4-(1,1-dimethylethyl)-2-ethoxyphenyl]-4,5-dihydrooxazole, Baroque, Borneo, Etoxazole, TetraSan, YI 5301, Yimanzuo, Zeal
IUPAC Name 4-(4-tert-butyl-2-ethoxyphenyl)-2-(2,6-difluorophenyl)-4,5-dihydro-1,3-oxazole
SMILES CCOc1cc(ccc1C2COC(=N2)c3c(F)cccc3F)C(C)(C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C21H23F2NO2/c1-5-25-18-11-13(21(2,3)4)9-10-14(18)17-12-26-20(24-17)19-15(22)7-6-8-16(19)23/h6-11,17H,5,12H2,1-4H3
Formula Weight 359.1697
Percent Purity >95%
Chemical Name or Material Etoxazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.