missing translation for 'onlineSavingsMsg'
Learn More

Etoxazole 10 μg/mL in Cyclohexane, Dr. Ehrenstorfer

Product Code. 30312400
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312400 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312400 Supplier Dr. Ehrenstorfer Supplier No. DREL13368000CY
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Etoxazole
CAS 153233-91-1
Analyte or Component Names Etoxazole
Shipping Condition Room Temperature
Molecular Formula C21 H23 F2 N O2
Concentration 10 μg/mL
Certifications/Compliance ISO/IEC 17025
Synonym Oxazole, 2-(2,6-difluorophenyl)-4-[4-(1,1-dimethylethyl)-2-ethoxyphenyl]-4,5-dihydro-, 2-(2,6-Difluorophenyl)-4-[4-(1,1-dimethylethyl)-2-ethoxyphenyl]-4,5-dihydrooxazole, Baroque, Borneo, Etoxazole, TetraSan, YI 5301, Yimanzuo, Zeal
InChI Formula InChI=1S/C21H23F2NO2/c1-5-25-18-11-13(21(2,3)4)9-10-14(18)17-12-26-20(24-17)19-15(22)7-6-8-16(19)23/h6-11,17H,5,12H2,1-4H3
Solution Type Single Solution
SMILES CCOc1cc(ccc1C2COC(=N2)c3c(F)cccc3F)C(C)(C)C
IUPAC Name 4-(4-tert-butyl-2-ethoxyphenyl)-2-(2,6-difluorophenyl)-4,5-dihydro-1,3-oxazole
Solvent or Matrix Cyclohexane
Industry Type Environmental Testing
Molecular Weight (g/mol) 359.41
Quantity 10 mL
Formula Weight 359.1697 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.