missing translation for 'onlineSavingsMsg'
Learn More

Etidronic Acid Disodium Salt, TRC

Product Code. 30137585
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137585 1 g 1g
30132489 5 g 5g
17970445 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137585 Supplier Toronto Research Chemicals Supplier No. TRCE9350001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7414-83-7
Molecular Formula C2 H6 O7 P2 . 2 Na
Molecular Weight (g/mol) 249.99
Synonym Etidronate disodium, Disodium dihydrogen (1-hydroxyethylidene)bisphosphonate
IUPAC Name disodium;hydroxy-[1-hydroxy-1-[hydroxy(oxido)phosphoryl]ethyl]phosphinate
SMILES [Na+].[Na+].CC(O)(P(=O)(O)[O-])P(=O)(O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H8O7P2.2Na/c1-2(3,10(4,5)6)11(7,8)9;;/h3H,1H3,(H2,4,5,6)(H2,7,8,9);;/q;2*+1/p-2
Formula Weight 249.9384
Percent Purity >95%
Chemical Name or Material Etidronate disodium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.