missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etidronate Disodium, Mikromol™
Description
Etidronate Disodium
Specifications
Specifications
| Chemical Name or Material | Etidronate disodium |
| CAS | 7414-83-7 |
| Analyte or Component Names | Etidronate disodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C2 H6 O7 P2 . 2 Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Etidronate disodium, Disodium dihydrogen (1-hydroxyethylidene)bisphosphonate |
| InChI Formula | InChI=1S/C2H8O7P2.2Na/c1-2(3,10(4,5)6)11(7,8)9;;/h3H,1H3,(H2,4,5,6)(H2,7,8,9);;/q;2*+1/p-2 |
| SMILES | [Na+].[Na+].CC(O)(P(=O)(O)[O-])P(=O)(O)[O-] |
| IUPAC Name | disodium;hydroxy-[1-hydroxy-1-[hydroxy(oxido)phosphoryl]ethyl]phosphinate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?