Learn More
Ethylene Terephthalate Cyclic Trimer, TRC
Product Code. 30006085
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006085 50 mg 50mg
30012076 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30006085 Supplier Toronto Research Chemicals Supplier No. TRCE91771550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7441-32-9
Molecular Formula C30H24O12
Molecular Weight (g/mol) 576.5
Synonym 3,6,13,16,23,26-Hexaoxatetracyclo[26.2.2.28,11.218,21]hexatriaconta-8,10,18,20,28,30,31,33,35-nonaene-2,7,12,17,22,27-hexone, Terephthalic acid, trimol. cyclic ethylene ester, Ethylene glycol, trimol. cyclic terephthalate, 1,4,11,14,21,24-Hexaoxatribenzo[abc,klm,uvw]cyclotriacontane-5,10,15,20,25,30-hexone, Cyclic ethylene terephthalate trimer, Cyclic tris(ethylene terephthalate), Cyclotris(ethylene glycol terephthalate), Ethylene glycol-terephthalic acid cyclic trimer, Ethylene terephthalate cyclic trimer, Ethylene terephthalate trimer, Tris ethylene terephthalate
SMILES O=C1OCCOC(=O)c2ccc(cc2)C(=O)OCCOC(=O)c3ccc(cc3)C(=O)OCCOC(=O)c4ccc1cc4
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C30H24O12/c31-25-19-1-2-20(4-3-19)26(32)38-15-16-40-28(34)22-9-11-24(12-10-22)30(36)42-18-17-41-29(35)23-7-5-21(6-8-23)27(33)39-14-13-37-25/h1-12H,13-18H2
Percent Purity >95%
Chemical Name or Material Ethylene Terephthalate Cyclic Trimer
Grade HPLC