Learn More
Ethylene Terephthalate Cyclic Tetramer, TRC
Product Code. 17978224
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17978224 2.5 mg 2.5mg
17977924 5 mg 5mg
17918514 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17978224 Supplier Toronto Research Chemicals Supplier No. TRCE9181902.5MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16104-96-4
Molecular Formula C40 H32 O16
Molecular Weight (g/mol) 768.672
Synonym 3,6,13,16,23,26,33,36-Octaoxapentacyclo[36.2.2.28,11.218,21.228,31]octatetraconta-8,10,18,20,28,30,38,40,41,43,45,47-dodecaene-2,7,12,17,22,27,32,37-octone, Terephthalic Acid, Tetramol. Cyclic Ethylene Ester, 3,6,13,16,23,26,33,36-Octaoxapentacyclo[36.2.2.28,11.218,21.228,31]octatetraconta-8,10,18,20,28,30,38,40,41,43,45,47-dodecaen-2,7,12,17,22,27,32,37-octone, Ethylene Glycol, Tetramol. Cyclic Terephthalate, Ethylene Terephthalate Cyclic Tetramer
SMILES O=C1OCCOC(=O)c2ccc(cc2)C(=O)OCCOC(=O)c3ccc(cc3)C(=O)OCCOC(=O)c4ccc(cc4)C(=O)OCCOC(=O)c5ccc1cc5
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C40H32O16/c41-33-25-1-2-26(4-3-25)34(42)50-19-20-52-36(44)28-9-11-30(12-10-28)38(46)54-23-24-56-40(48)32-15-13-31(14-16-32)39(47)55-22-21-53-37(45)29-7-5-27(6-8-29)35(43)51-18-17-49-33/h1-16H,17-24H2
Formula Weight 768.169
Chemical Name or Material Ethylene Terephthalate Cyclic Tetramer