Learn More
Ethylene Terephthalate Cyclic Dimer, TRC
Product Code. 30010994
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010994 10 mg 10mg
30006643 1 mg 1mg
30007955 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010994 Supplier Toronto Research Chemicals Supplier No. TRCE91817010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24388-68-9
Molecular Formula C20 H16 O8
Molecular Weight (g/mol) 384.3362
Synonym 3,6,13,16-Tetraoxatricyclo[16.2.2.28,11]tetracosa-8,10,18,20,21,23-hexaene-2,7,12,17-tetrone, Terephthalic Acid, Bimol. Cyclic Ethylene Ester, Ethylene Glycol, Bimol. Cyclic Terephthalate
SMILES O=C1OCCOC(=O)c2ccc(cc2)C(=O)OCCOC(=O)c3ccc1cc3
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C20H16O8/c21-17-13-1-2-14(4-3-13)18(22)26-11-12-28-20(24)16-7-5-15(6-8-16)19(23)27-10-9-25-17/h1-8H,9-12H2
Formula Weight 384.0845
Percent Purity >95%
Chemical Name or Material Ethylene Terephthalate Cyclic Dimer
Grade HPLC