missing translation for 'onlineSavingsMsg'
Learn More

Ethylamine hydrochloride, 99%

Product Code. 10380984 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10380984 2.5 kg 2.5kg
10398630 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10380984 Supplier Thermo Scientific Acros Supplier No. 214250025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 557-66-4 | C2H8ClN | 81.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-66-4
Molecular Formula C2H8ClN
Molecular Weight (g/mol) 81.54
InChI Key XWBDWHCCBGMXKG-UHFFFAOYSA-N
IUPAC Name ethanamine hydrochloride
SMILES Cl.CCN

Specifications

Melting Point 105.0°C to 108.0°C
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C2H5NH2·HCl
Quantity 2.5 kg
Beilstein 04, 91
Merck Index 15, 3817
Solubility Information Solubility aq. soln.: soluble. Other solubilities: freely soluble in alcohol, slightly soluble in chcl3 and acetone, practically insoluble in ether
Formula Weight 81.55
Chemical Name or Material Ethylamine hydrochloride
compliance-icons
Product Identifier
  • Ethylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.