accessibility menu, dialog, popup

UserName

CAS RN 557-66-4

HClH3CNH2

CAS RN 557-66-4

IUPAC Name: ethanamine hydrochloride
Synonyms: ethylamine, hydrochloride
Molecular Weight (g/mol): 81.54
Molecular Formula: C2H8ClN
InChi Key: XWBDWHCCBGMXKG-UHFFFAOYSA-N
SMILES: Cl.CCN
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)