Learn More
Ethyl-tert-butyl ether 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30318537
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318537 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30318537 Supplier Dr. Ehrenstorfer Supplier No. DREA13321300AL100
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl tert-Butyl Ether (ETBE)
CAS 637-92-3
Analyte or Component Names Ethyl tert-Butyl Ether (ETBE)
Shipping Condition Room Temperature
Molecular Formula C6 H14 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Propane, 2-ethoxy-2-methyl-, Ether, tert-butyl ethyl (6CI,7CI,8CI), 2-Ethoxy-2-methylpropane, 1,1-Dimethylethyl ethyl ether, Ethyl 1,1-dimethylethyl ether, Ethyl tert-butyl ether, Ethyl tert-butyl oxide, NSC 1069, tert-Butyl ethyl ether
InChI Formula InChI=1S/C6H14O/c1-5-7-6(2,3)4/h5H2,1-4H3
SMILES CCOC(C)(C)C
IUPAC Name 2-ethoxy-2-methylpropane
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 102.17
Quantity 1 mL
Formula Weight 102.1045 g/mol