Learn More
Ethyl N-(mesitylsulfonyl)oxyacetimidate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 38202-27-6 |
| Molecular Formula | C13H19NO4S |
| MDL Number | MFCD00009244 |
| Quantity | 25 g |
| SMILES | C/C(OCC)=N\OS(=O)(C1=C(C)C=C(C)C=C1C)=O |
| Molecular Weight (g/mol) | 285.36 |
| Percent Purity | 98% |
| Chemical Name or Material | Ethyl N-(mesitylsulfonyl)oxyacetimidate |