missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-m-Aminobenzoate, methanesulfonate salt, MP Biomedicals™

Product Code. 11453501 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
Unit Size:
100g
250g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453501 250 g 250g
11423501 5 g 5g
11433501 25 g 25g
11443501 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11453501 Supplier MP Biomedicals Supplier No. 0210310683
  • ONLINE EXCLUSIVE PRICE
    1045.00 EUR
    994.35 EUR / 250g
    Save 50.65 EUR
    4% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ethyl-m-Aminobenzoate is a derivative of an isomer of benzocaine

Chemical Identifiers

CAS 886-86-2
Molecular Formula C10H15NO5S
Molecular Weight (g/mol) 261.292
MDL Number MFCD00013176
InChI Key FQZJYWMRQDKBQN-UHFFFAOYSA-N
Synonym ethyl 3-aminobenzoate methanesulfonate, tricaine, tricaine methanesulfonate, finquel, metacaine, tricaine mesylate, 3-aminobenzoic acid ethyl ester methanesulfonate, ethyl m-aminobenzoate methanesulfonate, ethyl 3-aminobenzoate methanesulfonic acid salt, 3-aminobenzoic acid ethyl ester methanesulfonate salt
PubChem CID 261501
IUPAC Name ethyl 3-aminobenzoate;methanesulfonic acid
SMILES CCOC(=O)C1=CC(=CC=C1)N.CS(=O)(=O)O

Specifications

Color White
Density Ethyl-m-Aminobenzoate is mainly used as an anesthetic in fish,sharks,frogs
Assay Percent Range 0.993
Quantity 250 g
Formula Weight 261.3
Physical Form Powder
Chemical Name or Material Ethyl-m-Aminobenzoate, Methanesulfonate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.