missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Linoleate, TRC

Product Code. 30137756
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137756 100 g 100g
30137606 25 g 25g
30010366 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137756 Supplier Toronto Research Chemicals Supplier No. TRCE924300100G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-35-4
Molecular Formula C20 H36 O2
Molecular Weight (g/mol) 308.5
Synonym Ethyl Linoleate, (9Z,12Z)-Octadeca-9,12-dienoic Acid Ethyl Ester
IUPAC Name ethyl (9Z,12Z)-octadeca-9,12-dienoate
SMILES CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC

Specifications

Quantity 100 g
InChI Formula InChI=1S/C20H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h8-9,11-12H,3-7,10,13-19H2,1-2H3/b9-8-,12-11-
Formula Weight 308.2715
Chemical Name or Material Linoleic acid ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.