missing translation for 'onlineSavingsMsg'
Learn More

Ethyl α-Isopropylacetoacetate, Pract, MP Biomedicals

Product Code. 11469454 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469454 10 g 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11469454 Supplier MP Biomedicals Supplier No. 0522278525

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1522-46-9
Molecular Formula C9H16O3
MDL Number MFCD00042884
Quantity 10 g
Synonym ethyl 2-isopropylacetoacetate, 2-isopropylacetoacetic acid ethyl ester, ethyl 2-iso-propylacetoacetate, butanoic acid, 2-acetyl-3-methyl-, ethyl ester, ethyl alpha-isopropylacetoacetate, ethyl 2-acetyl-3-methylbutyrate, acetoacetic acid, 2-isopropyl-, ethyl ester, ethyl .alpha.-isopropylacetoacetate, ethyl isopropylaceto-acetate, acmc-1c35s
InChI Key DMIFFKCVURTPTG-UHFFFAOYNA-N
SMILES CCOC(=O)C(C(C)C)C(C)=O
IUPAC Name ethyl 2-acetyl-3-methylbutanoate
Molecular Weight (g/mol) 172.22
PubChem CID 95544
Formula Weight 172.23
Chemical Name or Material Ethyl-Alpha-Isopropylacetoacetate, Pract
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.