Learn More
Ethyl Diphenylphosphinite, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl Diphenylphosphinite |
| CAS | 719-80-2 |
| Molecular Formula | C14H15OP |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C14H15OP/c1-2-15-16(13-9-5-3-6-10-13)14-11-7-4-8-12-14/h3-12H,2H2,1H3 |
| SMILES | CCOP(c1ccccc1)c2ccccc2 |
| IUPAC Name | ethoxy(diphenyl)phosphane |
| Molecular Weight (g/mol) | 230.24 |
| Formula Weight | 230.0861 |