Learn More
Ethyl Cyanoacetate, TRC
Product Code. 30373097
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373097 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30373097 Supplier Toronto Research Chemicals Supplier No. TRCE905555100G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl Cyanoacetate
CAS 105-56-6
Molecular Formula C5H7NO2
Quantity 100 g
Synonym Cyanoacetic Acid Ethyl Ester, (Ethoxycarbonyl)acetonitrile, Cyanoacetic Acid Ethyl Ester, Cyanoacetic Ester, Ethyl 2-Cyanoacetate, Ethyl Cyanacetate, Ethyl Cyanoacetate, Ethyl Cyanoethanoate, Ethyl alpha-Cyanoacetate, Malonic Acid Ethyl Ester Nitrile, NSC 8844
InChI Formula InChI=1S/C5H7NO2/c1-2-8-5(7)3-4-6/h2-3H2,1H3
SMILES CCOC(=O)CC#N
IUPAC Name ethyl 2-cyanoacetate
Molecular Weight (g/mol) 113.11
Formula Weight 113.0477